Learn More
Y06036, MedChemExpress
MedChemExpress Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.
Brand:  Medchem Express HY-111502
Encompass_Preferred
 View more versions of this product
Product Code. 16463106
  • POA / 25mg
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.
Specifications
Specifications
Y06036
Light Yellow
C16H15BrN2O5S
DMSO : 130 mg/mL (304.26 mM; Need ultrasonic)
427.27
98.96%
Solid
1832671-96-1
Research
25 mg
BrC1=CC(S(=O)(NC2=C(OC)C=C(ON=C3C)C3=C2)=O)=C(OC)C=C1
427.27
Research
Neuroscience-Neuromodulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Y06036, MedChemExpress