Learn More
Y06036, MedChemExpress
MedChemExpress Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.
Brand: Medchem Express HY-111502
This item is not returnable.
View return policy
Description
Y06036 is a potent and selective BET inhibitor, which binds to the BRD4(1) bromodomain with Kd value of 82 nM. Antitumor activity.Specifications
Y06036 | |
Light Yellow | |
C16H15BrN2O5S | |
DMSO : 130 mg/mL (304.26 mM; Need ultrasonic) | |
427.27 | |
98.96% | |
Solid |
1832671-96-1 | |
Research | |
25 mg | |
BrC1=CC(S(=O)(NC2=C(OC)C=C(ON=C3C)C3=C2)=O)=C(OC)C=C1 | |
427.27 | |
Research | |
Neuroscience-Neuromodulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Y06036, MedChemExpress