Learn More
B Braun™ Yasargil™ Aneurysm Clips
Intended for occlusion of cerebral aneurysms in either a temporary or permanent manner.
322.00 €
Specifications
Material Titanium
View More Specs
Products 1
Product Code Brand Length (Metric) Product Type Tip Style Price Quantity & Availability  
Product Code Brand Length (Metric) Product Type Tip Style Price Quantity & Availability  
15894428
View Documents
B Braun™
FE769K
11.0 mm (Blade) Standard, Temporary Phynox 7.8 mm opening This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Smooth surface prevents damage to tissue
  • Deep, symmetrical pyramidal stamping of both blades doubles the contact surface area and counteracts the slip effect
  • Flat design with a broad contact surface enables smooth opening and closing as well as stability
  • Lid design of one-piece construction for easier maintenance and lower repair costs
Specifications
Specifications
Titanium
Videos
SDS
Documents
Documents
Product Certifications