Learn More
LNI Swissgas ZA TOTAL C 1.5
Zero air generator for HC/carbon dioxide/moisture removal with integrated compressor.
Brand:  LNI Swissgas 6920.25.215
Product Code. 17832099
  • 8400.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
  • Zero air generator 1.5 L/min HC/carbon dioxide/moisture removal with integrated compressor.
Specifications
Specifications
100/115/220/230 V
Zero Air Gas Generator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LNI Swissgas ZA TOTAL C 1.5 > LNI Swissgas Zero Air Generator