Learn More
R&L Enterprises™ Zinc Plated Stand Rods
Product Code. p-8012548
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
Diameter (Metric) Rod:
10 mm
12.5 mm
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 12956806
Please to purchase this item. Need a web account? Register with us today!
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Zinc Plated Rods are manufactured from mild steel.
  • Threaded
  • Available for a range of diameters and lengths
  • Length of rod does not include the thread
Specifications
Material Zinc Plated Mild Steel
Length (Metric) 750 mm
Diameter (Metric) Rod 12.5 mm
Product Type Rod