missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc solution for ICP 10g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
103.00 € - 279.00 €
Specifications
Concentration | 10±0.02g/L |
---|---|
Type | Mass Spectrometry Standard |
Analyte or Component Names | Zn |
Solvent or Matrix | HNO3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12927243
|
Fisher Chemical
S 369 100 |
100 mL | Bottle |
103.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12937243
|
Fisher Chemical
S 369 250 |
250 mL | Bottle |
279.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12947243
|
Fisher Chemical
S 369 500 |
500 mL | Bottle |
235.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10±0.02g/L | |
Mass Spectrometry Standard | |
Zn | |
HNO3 |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Zinc (Zn) 10000mg/l in HNO3 5%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title