Learn More
RETSCH Zirconium Oxide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Zirconium Oxide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand:  RETSCH 01.462.0227
 View more versions of this product
Product Code. 10161002
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
View alternative products »
Explore more special offers
Description
Description
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Specifications
Comfort Grinding Jar
500 mL
For PM 100, PM 400
Zirconium Oxide
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products