Learn More
RETSCH Zirconium Oxide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Zirconium Oxide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand: RETSCH
This item is not returnable.
View return policy
Description
- Available in a variety of sizes
- Optional aeration covers
- Pressure tight safety closure devices
Specifications
Comfort Grinding Jar | |
125 mL | |
For PM 100, PM 200, PM 400 and S 100 | |
Zirconium Oxide |