accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1026582-88-6

H3CCH3NNH2NNH2OH3COCH3

CAS RN 1026582-88-6

IUPAC Name: 5-{[4,5-dimethoxy-2-(propan-2-yl)phenyl]methyl}pyrimidine-2,4-diamine
Synonyms: 5-[(2-isopropyl-4,5-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol): 302.38
Molecular Formula: C16H22N4O2
InChi Key: PYNPWUIBJMVRIG-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C(C)C)C(CC2=CN=C(N)N=C2N)=C1
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.