Learn More
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride, 97%, Thermo Scientific Chemicals
186.00 € - 694.00 €
Chemical Identifiers
CAS | 1171331-39-7 |
---|---|
Molecular Formula | C4H8ClF3N2O |
Molecular Weight (g/mol) | 192.566 |
MDL Number | MFCD09944516 |
InChI Key | DBNFKWRZLGVLSH-UHFFFAOYSA-N |
Synonym | 2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl |
PubChem CID | 42913698 |
IUPAC Name | 2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride |
SMILES | C(C(=O)NCC(F)(F)F)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15490636
|
Thermo Scientific Alfa Aesar
H53398.03 |
1 g |
186.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400646
|
Thermo Scientific Alfa Aesar
H53398.06 |
5 g |
694.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reactant in the synthesis of isoxazoline indolizine amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis of isoxazoline indolizine amides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
1171331-39-7 | |
192.566 | |
DBNFKWRZLGVLSH-UHFFFAOYSA-N | |
42913698 | |
C(C(=O)NCC(F)(F)F)N.Cl |
C4H8ClF3N2O | |
MFCD09944516 | |
2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl | |
2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride |
Specifications
1171331-39-7 | |
H2NCH2(CO)NHCH2CF3·HCl | |
2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl | |
C(C(=O)NCC(F)(F)F)N.Cl | |
192.566 | |
192.57 | |
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride |
C4H8ClF3N2O | |
MFCD09944516 | |
DBNFKWRZLGVLSH-UHFFFAOYSA-N | |
2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride | |
42913698 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only