Learn More
3,4-Dichlorobenzotrifluoride, 97%, Thermo Scientific™
15.88 € - 127.00 €
Chemical Identifiers
CAS | 328-84-7 |
---|---|
Molecular Formula | C7H3Cl2F3 |
Molecular Weight (g/mol) | 214.996 |
MDL Number | MFCD00000555 |
InChI Key | XILPLWOGHPSJBK-UHFFFAOYSA-N |
Synonym | 3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte |
PubChem CID | 9481 |
IUPAC Name | 1,2-dichloro-4-(trifluoromethyl)benzene |
SMILES | C1=CC(=C(C=C1C(F)(F)F)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11457407
|
Thermo Scientific Alfa Aesar
A15098.09 |
10 g | N/A | ||||||
11467407
|
Thermo Scientific Alfa Aesar
A15098.18 |
50 g | N/A | ||||||
11319539
|
Thermo Scientific Alfa Aesar
A15098.30 |
250 g | N/A | ||||||
11329539
|
Thermo Scientific Alfa Aesar
A15098.36 |
500 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
328-84-7 | |
214.996 | |
XILPLWOGHPSJBK-UHFFFAOYSA-N | |
9481 | |
C1=CC(=C(C=C1C(F)(F)F)Cl)Cl |
C7H3Cl2F3 | |
MFCD00000555 | |
3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte | |
1,2-dichloro-4-(trifluoromethyl)benzene |
Specifications
328-84-7 | |
1.479 | |
65°C (149°F) | |
1.475 | |
UN1760 | |
3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte | |
C1=CC(=C(C=C1C(F)(F)F)Cl)Cl | |
214.996 | |
215 | |
3,4-Dichlorobenzotrifluoride |
-12°C | |
173°C to 174°C | |
C7H3Cl2F3 | |
MFCD00000555 | |
1950151 | |
XILPLWOGHPSJBK-UHFFFAOYSA-N | |
1,2-dichloro-4-(trifluoromethyl)benzene | |
9481 | |
97% |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 206-337-1
missing translation for 'rtecsNumber' : CZ5527510
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.