missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3730 DNA Analyzer 48-Capillary Array, 36 cm
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 array
Unit Size:
Each
Description
Includes
Each array contains 48 capillaries (36cm in length)
- Usage per array: 300 runs (with use of Applied Biosystems reagents)
Order Info
Each
Specifications
Specifications
| Capillary Length | 36 cm |
| Content And Storage | Each array contains 48 capillaries (36 cm in length) for use with the Applied Biosystems 3730 DNA Analyzer. We guarantee the performance of the 3730 Capillary arrays up to 300 runs and within 1 year of purchase. |
| Format | Capillary Array |
| For Use With (Equipment) | 3730 DNA Analyzer |
| For Use With (Application) | Sequencing |
| Number of Capillaries | 48 Capillaries |
| Product Type | 3730 DNA Analyzer 48-Capillary Array |
| Quantity | 1 array |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction