Learn More
Memmert™ Additional Stainless Steel Grid for UN260plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ UN260plusE29766
This item is not returnable.
View return policy
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
UN260plus Oven | |
Reinforced Grid | |
Stainless Steel | |
60 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Additional Stainless Steel Grid for UN260plus, Reinforced Chamber Option > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction