Learn More
Bifenazate, MedChemExpress
MedChemExpress Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.
Brand: Medchem Express HY-119687
Description
Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.Specifications
Bifenazate | |
Yellow | |
C17H20N2O3 | |
DMSO : ≥ 250 mg/mL (832.36 mM) | |
300.35 | |
99.65% | |
Solid |
149877-41-8 | |
Research | |
1 g | |
COC1=CC=C(C2=CC=CC=C2)C=C1NNC(OC(C)C)=O | |
300.35 | |
Research | |
Neuroscience-Neuromodulation |