Learn More
Amylin Antibody (R10/99), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Bio-Techne NB600-713-0.25ML
Description
Amylin Monoclonal specifically detects Amylin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Amylin | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant with 0.09% Sodium Azide | |
AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin) | |
Mouse | |
Tissue culture supernatant | |
RUO | |
Primary | |
The is of high affinity, and stains islet amyloid peptide in intra-cellular insulin granules, intra-cellular amyloid and extra-cellular islet amyloid. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
R10/99 | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
P10997 | |
IAPP | |
Synthetic peptide: CATQRLANFLV, corresponding to internal sequence amino acids 40-50 of Human Amylin coupled to tuberculin. | |
0.25 mL | |
Angiogenesis, Apoptosis, Cancer | |
3375 | |
Human | |
Supernatant |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only