missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amylin Antibody (R10/99), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
209.00 € - 477.00 €
Specifications
Antigen | Amylin |
---|---|
Clone | R10/99 |
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18047233
|
Bio-Techne
NB600-713-0.25ML |
0.25 mL |
209.00 €
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18205645
|
Bio-Techne
NB600-713 |
1 mL |
477.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amylin Monoclonal specifically detects Amylin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Amylin | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin) | |
IAPP | |
IgG1 | |
Tissue culture supernatant | |
The is of high affinity, and stains islet amyloid peptide in intra-cellular insulin granules, intra-cellular amyloid and extra-cellular islet amyloid. |
R10/99 | |
Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
Unconjugated | |
Mouse | |
Angiogenesis, Apoptosis, Cancer | |
P10997 | |
3375 | |
Synthetic peptide: CATQRLANFLV, corresponding to internal sequence amino acids 40-50 of Human Amylin coupled to tuberculin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Amylin Antibody (R10/99), Novus Biologicals™