Learn More
Anti-Bumping Granules, Extra Pure, Fused Alumina, SLR, Fisher Chemical™
Added to liquids to make them boil more calmly.
Brand: Fisher Chemical A/7770/50
54.99 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Anti-bumping Granules | |
Fused Alumina | |
250 g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Anti-Bumping Granules, Extra Pure, Fused Alumina, SLR, Fisher Chemical™ > 250g, Plastic powder jar
Spot an opportunity for improvement?Share a Content Correction