Learn More
Thermo Scientific Chemicals Boiling Ston
For analytical work
Brand: Thermo Scientific Chemicals 386202000
57.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Stone | |
200 g |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Boiling stones
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466948 | Certificate of Analysis | 18/10/2024 | 386200500,386202000 |
A0463527 | Certificate of Analysis | 12/06/2024 | 386200500,386202000 |
A0461196 | Certificate of Analysis | 05/06/2024 | 386200500,386202000 |
A0455436 | Certificate of Analysis | 25/08/2023 | 386200500,386202000 |
A0447304 | Certificate of Analysis | 30/12/2022 | 386200500,386202000 |