Learn More
Karl Hecht™ Assisent™ Polyethylene Pasteur Pipets
Ideal for handling liquids safely in controlled, reproducible drop sizes. Karl Hecht™ Assistent™ Polyethylene Pasteur Pipets are applicable for all types of laboratory tests and analyses.
Brand: Karl Hecht™ 40569003
This item is not returnable.
View return policy
Description
- Manufactured from high quality, chemically inert, non-toxic, low-density polyethylene
- Excellent for transferring samples to sample cups and for drop counting
- Disposable
Specifications
Polyethylene | |
3 mL | |
Pasteur Pipet | |
Single-use |