Learn More
Karl Hecht™ Assisent™ Polyethylene Pasteur Pipets
Ideal for handling liquids safely in controlled, reproducible drop sizes. Karl Hecht™ Assistent™ Polyethylene Pasteur Pipets are applicable for all types of laboratory tests and analyses.
22.00 € - 34.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10401132
|
Karl Hecht™
40569002 |
1 mL |
22.00 €
Pack of 500 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
||||||
10247661
|
Karl Hecht™
40569003 |
3 mL |
34.00 €
Pack of 500 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||||
Description
- Manufactured from high quality, chemically inert, non-toxic, low-density polyethylene
- Excellent for transferring samples to sample cups and for drop counting
- Disposable
Specifications
Polyethylene | |
Pasteur Pipet | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Hecht™ Assisent™ Polyethylene Pasteur Pipets