Learn More
BAY-545, MedChemExpress
MedChemExpress BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.
Brand: Medchem Express HY-111767
Description
BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.Specifications
BAY-545 | |
1699717-32-2 | |
10 mg | |
C18H22F3N3O4S | |
DMSO : ≥ 150 mg/mL (346.06 mM) | |
433.45 | |
95.88% |
Solid | |
White | |
Research | |
COVID-19-immunoregulation | |
CCN1C(N(CCC(F)(F)F)C2=C(C(C)=C(C(N3CCC(O)CC3)=O)S2)C1=O)=O | |
433.45 | |
Research |