Learn More
BAY-545, MedChemExpress
MedChemExpress BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.

488.70 € - 2718.90 €
Specifications
Chemical Name or Material | BAY-545 |
---|---|
Physical Form | Solid |
CAS | 1699717-32-2 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490827
![]() |
Medchem Express
HY-111767 |
50 mg |
1888.65 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16457237
![]() |
Medchem Express
HY-111767 |
5 mg |
488.70 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16425386
![]() |
Medchem Express
HY-111767 |
100 mg |
2718.90 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424528
![]() |
Medchem Express
HY-111767 |
10 mg |
731.70 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.Specifications
BAY-545 | |
1699717-32-2 | |
Research | |
COVID-19-immunoregulation | |
CCN1C(N(CCC(F)(F)F)C2=C(C(C)=C(C(N3CCC(O)CC3)=O)S2)C1=O)=O | |
433.45 | |
Research |
Solid | |
White | |
C18H22F3N3O4S | |
DMSO : ≥ 150 mg/mL (346.06 mM) | |
433.45 | |
95.88% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.