Learn More
BAY-545, MedChemExpress
MedChemExpress BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.
Brand:  Medchem Express HY-111767
Encompass_Preferred
 View more versions of this product
Product Code. 16424528
  • 731.70 € / 10mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.
Specifications
Specifications
BAY-545
1699717-32-2
10 mg
C18H22F3N3O4S
DMSO : ≥ 150 mg/mL (346.06 mM)
433.45
95.88%
Solid
White
Research
COVID-19-immunoregulation
CCN1C(N(CCC(F)(F)F)C2=C(C(C)=C(C(N3CCC(O)CC3)=O)S2)C1=O)=O
433.45
Research
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BAY-545, MedChemExpress