Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefotaxime/Clavulanic Acid
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand: BD 231752
This item is not returnable.
View return policy
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Cefotaxime/Clavulanic Acid | |
10μg | |
Disc | |
10 Pack |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Disc Cefotaxime/Clavulanic Ac 10 Ea
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium [6R-[6]]-3-(acetoxymethyl)-7-[(2-aminothiazol-4-yl)(methoxyimino)acetamido]-8-oxo-5-th
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction