missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefotaxime/Clavulanic Acid
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
37.15 € - 182.00 €
Specifications
Antimicrobial Agents | Cefotaxime/Clavulanic Acid |
---|---|
Concentration | 10μg |
Packaging Type | Disc |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17362441
|
BD
231751 |
1 Pack |
37.15 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
17372441
|
BD
231752 |
10 Pack |
182.00 €
Pack of 10 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||||
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Cefotaxime/Clavulanic Acid | |
10μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 365 Days
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Cefotaxime/Clavulanic Acid 30/10Mcg
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium [6R-[6]]-3-(acetoxymethyl)-7-[(2-aminothiazol-4-yl)(methoxyimino)acetamido]-8-oxo-5-th
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title