Learn More
Boekel Scientific™ Platform for Microprocessor Controlled Orbital Shaker
GREENER_CHOICE
This is an interchangeable platform for Boekel Orbital Shakers (Cat. No. 15-600-317).
Brand:  Boekel Scientific™ 270290
 View more versions of this product
Product Code. 12673555
  • 203.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Platform with Holders for Flasks and Beakers
  • interchangeable Locking Design
Specifications
Specifications
2.5 kg
Platform
250 mm
190 mm
6 x 250 mL Flasks, 6 x 400 mL Beakers
9.8 in.
7.5 in.
Boekel Microprocessor Controlled Orbital Shaker