Learn More
Bohlender™ BOLA™ Gas Distributors
Gas Distributors for Scrubber Adapters for Bottles and Scrubber Bottles Vitrum.
Brand:  Bohlender™ N910-02
 View more versions of this product
Product Code. 17157554
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Made of PTFE
  • Suitable for BOLA™ Scrubber Adaptors and Scrubber Bottles Vitrum GL 45
  • Cat. No. N 1660-14, N 1662-14, N 1662-24
Specifications
Specifications
BOLA™ Scrubber Adaptors GLS 80 and Scrubber Bottles Vitrum GL 45 (Cat. No. N 1660-24, N 1662-34, N 1662-44)
PTFE
5 Pk.