Learn More
Bohlender™ BOLA™ Gas Distributors
Gas Distributors for Scrubber Adapters for Bottles and Scrubber Bottles Vitrum.
47.00 € - 187.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17147554
|
Bohlender™
N910-01 |
BOLA Scrubber Adaptors GL 45 and Scrubber Bottles Vitrum GL 45 (Cat. No. N 1660-14, N 1662-14, N 1662-24) | N/A | ||||||
17834034
|
Bohlender™
N910-01 |
BOLA Scrubber Adaptors GL 45 and Scrubber Bottles Vitrum GL 45 (Cat. No. N 1660-14, N 1662-14, N 1662-24) |
47.00 €
Pack of 5 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
17157554
|
Bohlender™
N910-02 |
BOLA™ Scrubber Adaptors GLS 80 and Scrubber Bottles Vitrum GL 45 (Cat. No. N 1660-24, N 1662-34, N 1662-44) | N/A | ||||||
17814034
|
Bohlender™
N910-02 |
BOLA Scrubber Adaptors GLS 80 and Scrubber Bottles Vitrum GL 45 (Cat. No. N 1660-24, N 1662-34, N 1662-44) |
138.00 €
Pack of 5 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
- Made of PTFE
- Suitable for BOLA™ Scrubber Adaptors and Scrubber Bottles Vitrum GL 45
- Cat. No. N 1660-14, N 1662-14, N 1662-24
Specifications
PTFE | |
5 Pk. |