Learn More
Brady™ BBP™ Temperature Label Stock
BBP™ Cold Temperature Label Stock (B-549) is a flexible, dimensionally stable, pressure sensitive polyester tape perfect for use in any cold storage warehouse, cooler or freezer.
Brand:  Brady™ 117860
 View more versions of this product
Product Code. 11706375
  • 503.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Special adhesive can be applied to extremely cold surfaces (-10°F) and withstands temperatures as low as -40°F
  • Tape can be thermal transfer printed
  • Withstands mild chemicals, oil and water
  • Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Specifications
Specifications
B30C-4000-549-WT
Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
For general identification, ideal for cold storage warehouses, coolers, freezers and short-term winter outdoor use
20 Gardner Units
Up to 1 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +100°C
0.003 in.
100 ft.
4 in.
Cold Temperature Application Tape for BBP3x Printers
White
Label
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 212°F
35 lb./in.
0.076 mm
30.48 m
101 mm