Learn More
Brady™ BBP™ Temperature Label Stock
BBP™ Cold Temperature Label Stock (B-549) is a flexible, dimensionally stable, pressure sensitive polyester tape perfect for use in any cold storage warehouse, cooler or freezer.
Brand:  Brady™ 117865
 View more versions of this product
Product Code. 11756375
  • 238.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Special adhesive can be applied to extremely cold surfaces (-10°F) and withstands temperatures as low as -40°F
  • Tape can be thermal transfer printed
  • Withstands mild chemicals, oil and water
  • Service temperature of -40°F to 212°F (-40°C to 100°C)
Specifications
Specifications
B30C-1125-549-YL
Yellow
Label
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Up to 1 Years
-40 to 212°F
35 lb./in.
100 ft.
1.12 in.
Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
For general identification, ideal for cold storage warehouses, coolers, freezers and short-term winter outdoor use
20 Gardner Units
1 Roll
-40°C to +100°C
0.003 in.
30.48 m