Learn More
CD14 Monoclonal Antibody (TuK4), Pacific Blue™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1428
Description
Our Pacific Blue™ dye is optimally excited by the violet laser and conjugates of this dye are strongly fluorescent even at neutral pH. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.
Specifications
CD14 | |
Monoclonal | |
Pacific Blue | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
purified | |
ASR | |
929 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
TuK4 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P08571 | |
Cd14 | |
Human CD14 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.