missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, PE, Clone: Hit3a, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB7-PE-25Tests
This item is not returnable.
View return policy
Specifications
CD3 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD3E | |
Protein A/G Purified | |
RUO | |
916 | |
IgG2a κ |
Flow Cytometry | |
Hit3a | |
PE | |
NP_000724 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD3 Mouse, PE, Clone: Hit3a, enQuireBio™ > 25 Tests
Spot an opportunity for improvement?Share a Content Correction