missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, PE, Clone: Hit3a, enQuireBio™
Mouse Monoclonal Antibody
111.00 € - 187.00 €
Specifications
Antigen | CD3 |
---|---|
Clone | Hit3a |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952378
|
EnquireBio
QAB7-PE-25Tests |
25 Tests |
111.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942378
|
EnquireBio
QAB7-PE-100Tests |
100 Tests |
187.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD3 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
916 | |
IgG2a κ | |
Protein A/G Purified |
Hit3a | |
Flow Cytometry | |
PE | |
RUO | |
NP_000724 | |
CD3E | |
Primary | |
Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD3 Mouse, PE, Clone: Hit3a, enQuireBio™