missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD357 / GITR Rat, APC, Clone: DTA-1, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB72-APC-25ug
This item is not returnable.
View return policy
Specifications
CD357 / GITR | |
Monoclonal | |
0.5mg/mL | |
10mM Tris, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH8 | |
Tnfrsf18 | |
Protein A/G Purified | |
RUO | |
21936 | |
IgG2b |
Flow Cytometry | |
DTA-1 | |
APC | |
NP_033426 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |