missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD357 / GITR Rat, APC, Clone: DTA-1, enQuireBio™
Rat Monoclonal Antibody
162.00 € - 377.00 €
Specifications
Antigen | CD357 / GITR |
---|---|
Clone | DTA-1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952408
|
EnquireBio
QAB72-APC-25ug |
25 μg |
162.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942408
|
EnquireBio
QAB72-APC-100ug |
100 μg |
377.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD357 / GITR | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10mM Tris, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH8 | |
21936 | |
IgG2b | |
Protein A/G Purified |
DTA-1 | |
Flow Cytometry | |
APC | |
RUO | |
NP_033426 | |
Tnfrsf18 | |
Primary | |
Store at 2-4°C. |