Learn More
Cellpath CellSafe+ Blue Biopsy Capsules
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads which are proving to be unsatisfactory and time consuming. CellPath CellSafe+ Blue Biopsy Capsules features new blue mesh that provides better contrast between the biopsy and the CellSafe.
Brand: Cellpath EBE-0301-02A
This item is not returnable.
View return policy
Description
- Features two hinged interlocking frames with an integral nylon mesh
- Adjustable to biopsy specimen size
- Hinged in either direction and closed to encapsulate the tissue
- New Blue Mesh provides better contrast between the biopsy and the CellSafe
- X–Ray transparent mesh for breast work
- Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
- Zero Tissue damage or artefacts associated with foam pads
- Substantial time savings compared to folding biopsy in tissue paper
Specifications
100/Pk. | |
Blue |