Learn More
Cellpath CellSafe+ Blue Biopsy Capsules
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads which are proving to be unsatisfactory and time consuming. CellPath CellSafe+ Blue Biopsy Capsules features new blue mesh that provides better contrast between the biopsy and the CellSafe.
108.00 €
Specifications
Color Blue
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15158886
View Documents
Cellpath
EBE-0301-02A
100/Pk.
108.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Features two hinged interlocking frames with an integral nylon mesh
  • Adjustable to biopsy specimen size
  • Hinged in either direction and closed to encapsulate the tissue
  • New Blue Mesh provides better contrast between the biopsy and the CellSafe
  • X–Ray transparent mesh for breast work
  • Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
  • Zero Tissue damage or artefacts associated with foam pads
  • Substantial time savings compared to folding biopsy in tissue paper
Specifications
Specifications
Blue
Videos
SDS
Documents
Documents
Product Certifications