Learn More
Cephalosporin C zinc salt, MedChemExpress
MedChemExpress Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.
Brand:  Medchem Express HY-B1299A
Encompass_Preferred
 View more versions of this product
Product Code. 16464577
  • 238.95 € / 50mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.
Specifications
Specifications
Cephalosporin C zinc salt
Off-White
C16H19N3O8SZn
DMSO : 5 mg/mL (10.44 mM; ultrasonic and adjust pH to 3 with 1M HCl)
478.78
98.0%
Solid
59143-60-1
Research
50 mg
CC(OCC1=C(C([O-])=O)N(C([C@H]2NC(CCC[C@@H](N)C([O-])=O)=O)=O)[C@]2([H])SC1)=O.[Zn+2]
478.78
Research
COVID-19-immunoregulation