missing translation for 'onlineSavingsMsg'
Learn More

Cephalosporin C zinc salt, MedChemExpress

MedChemExpress Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.

106.65 € - 371.25 €

Specifications

CAS 59143-60-1
Molecular Formula C16H19N3O8SZn
Molecular Weight (g/mol) 478.78
Percent Purity 98.0%
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16498688
Encompass_Preferred
View Documents
Medchem Express
HY-B1299A
10 mg
106.65 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16464577
Encompass_Preferred
View Documents
Medchem Express
HY-B1299A
50 mg
238.95 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16418726
Encompass_Preferred
View Documents
Medchem Express
HY-B1299A
100 mg
371.25 €
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.
Specifications

Specifications

59143-60-1
C16H19N3O8SZn
478.78
98.0%
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only