Learn More
Witeg™ Class AS Graduated Volumetric Pipettes
Tempered delivery tips offering a wear resistance which is up to 7 times better than that of conventional pipette delivery tips.
4.45 € - 10.00 €
Specifications
Certifications/Compliance Conformity certified; DIN EN ISO 385
Class AS
Material Glass
Graduated Yes
Graduation Color Amber
View More Specs
Products 2
Product Code Brand Volume (Metric) Graduations Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Color Price Quantity & Availability  
10674392
View Documents
Witeg™
3 510 011
0.1 mL 0.001 mL divisions Green This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10214061
View Documents
Witeg™
3 510 810
20 mL 0.1 mL divisions Yellow
10.00 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Conformity certified
  • Main point ring graduation
  • With ISO color code
  • Acc. to DIN ENO ISO 385
  • With amber stain graduations and inscriptions
  • Sizes 5 mL and up with construction in the suction to hold cotton plug
  • Waiting time of 5 seconds
  • For complete and swift delivery
Specifications
Specifications
Conformity certified; DIN EN ISO 385
Glass
Amber
Tempered Delivery
AS
Yes
Main point ring
Graduated Pipet
Videos
SDS
Documents
Documents
Product Certifications