Learn More
COD Standard, 500mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Concentration | 500 mg/L |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
COD50024G1 | Certificate of Analysis | 29/01/2025 | COD500 |
COD50023G1 | Certificate of Analysis | 23/07/2024 | COD500 |
COD50024G1 | Certificate of Analysis | 16/07/2024 | COD500 |
COD50023C1 | Certificate of Analysis | 06/04/2023 | COD500 |
COD50022E1 | Certificate of Analysis | 16/11/2022 | COD500 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COD Standard, 500mg/L, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction