missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CXCL9 (Human) Recombinant Protein
Human CXCL9 full-length ORF ( AAH63122, 23 a.a. - 125 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | AAH63122 |
---|---|
Gene ID (Entrez) | 4283 |
Name | chemokine (C-X-C motif) ligand 9 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137831
|
Abnova™
H00004283-P01.10ug |
10 μg |
335.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16147831
|
Abnova™
H00004283-P01.25ug |
25 μg |
508.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sequence: TPVVRKGRCSCISTNQGTIHLQSLKDLKQFAPSPSCEKIEIIATLKNGVQTCLNPDSADVKELIKKWEKQVSQKKKQKNGKKHQKKKVLKVRKSQRSRQKKTT
Specifications
AAH63122 | |
chemokine (C-X-C motif) ligand 9 | |
125% SDS-PAGE Stained with Coomassie Blue | |
CMK, Humig, MIG, SCYB9, crg-10 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4283 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CXCL9 | |
GST |
Safety and Handling
missing translation for 'shelfLife' : Best use within three months from the date of receipt of this protein