missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Mouse, Clone: KRT10/844 + KRT10/1275, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3858-MSM6-P0
This item is not returnable.
View return policy
Specifications
Cytokeratin 10 | |
Cytoplasmic | |
KRT10/844 + KRT10/1275 | |
Unconjugated | |
NP_000412 | |
Mouse | |
56.5kDa | |
Protein A/G Purified | |
RUO | |
3858 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
KRT10 | |
Recombinant human KRT10 protein | |
A431, HeLa, MCF7 cells. Esophagus or tonsil. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |