missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Mouse, Clone: KRT10/844 + KRT10/1275, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 510.00 €
Specifications
Antigen | Cytokeratin 10 |
---|---|
Clone | KRT10/844 + KRT10/1275 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940358
|
EnquireBio
3858-MSM6-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15950358
|
EnquireBio
3858-MSM6-P1 |
100 μg |
510.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Cytokeratin 10 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
3858 | |
Recombinant human KRT10 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
56.5kDa |
KRT10/844 + KRT10/1275 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_000412 | |
KRT10 | |
IgG1 κ | |
Protein A/G Purified | |
Cytoplasmic | |
A431, HeLa, MCF7 cells. Esophagus or tonsil. |