missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Aspartic acid, 99%
27.80 € - 267.00 €
Chemical Identifiers
CAS | 1783-96-6 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.103 |
MDL Number | MFCD00063081 |
InChI Key | CKLJMWTZIZZHCS-UWTATZPHSA-N |
Synonym | d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d |
PubChem CID | 83887 |
ChEBI | CHEBI:17364 |
IUPAC Name | (2R)-2-aminobutanedioic acid |
SMILES | C(C(C(=O)O)N)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494677
|
Thermo Scientific Chemicals
B21184.06 |
5 g |
27.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404687
|
Thermo Scientific Chemicals
B21184.14 |
25 g |
85.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414687
|
Thermo Scientific Chemicals
B21184.22 |
100 g |
267.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Aspartic acid modulates melatonin synthesis in the pineal gland.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Aspartic acid modulates melatonin synthesis in the pineal gland.
Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
1783-96-6 | |
133.103 | |
CKLJMWTZIZZHCS-UWTATZPHSA-N | |
83887 | |
(2R)-2-aminobutanedioic acid |
C4H7NO4 | |
MFCD00063081 | |
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d | |
CHEBI:17364 | |
C(C(C(=O)O)N)C(=O)O |
Specifications
>300°C (decomposition) | |
C4H7NO4 | |
1723529 | |
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d | |
CKLJMWTZIZZHCS-UWTATZPHSA-N | |
(2R)-2-aminobutanedioic acid | |
83887 | |
133.1 | |
1.66 | |
D-Aspartic acid |
1783-96-6 | |
MFCD00063081 | |
14,840 | |
Slightly soluble in water,Soluble in 1N ammonium hydroxide (100mg/ml),and dilute HCl. Insoluble in ether,and alcohol. | |
C(C(C(=O)O)N)C(=O)O | |
133.103 | |
CHEBI:17364 | |
99% | |
−25° (c=1 in 1N HCl) |
Safety and Handling
missing translation for 'einecsNumber' : 217-234-6
missing translation for 'rtecsNumber' : CI9097500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only