missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glutamic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
21.40 € - 265.00 €
Chemical Identifiers
CAS | 27025-25-8 |
---|---|
Molecular Formula | C7H14ClNO4 |
Molecular Weight (g/mol) | 211.642 |
MDL Number | MFCD00235872 |
InChI Key | MFUPLHQOVIUESQ-NUBCRITNSA-N |
Synonym | dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride |
PubChem CID | 12917567 |
IUPAC Name | dimethyl (2R)-2-aminopentanedioate;hydrochloride |
SMILES | COC(=O)CCC(C(=O)OC)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458708
|
Thermo Scientific Alfa Aesar
H62132.03 |
1 g |
21.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15468708
|
Thermo Scientific Alfa Aesar
H62132.06 |
5 g |
57.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15437076
|
Thermo Scientific Alfa Aesar
H62132.14 |
25 g |
265.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
27025-25-8 | |
211.642 | |
MFUPLHQOVIUESQ-NUBCRITNSA-N | |
12917567 | |
COC(=O)CCC(C(=O)OC)N.Cl |
C7H14ClNO4 | |
MFCD00235872 | |
dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
dimethyl (2R)-2-aminopentanedioate;hydrochloride |
Spezifikation
91°C | |
C7H14ClNO4 | |
dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
COC(=O)CCC(C(=O)OC)N.Cl | |
211.642 | |
211.64 | |
−26° (c=5 in Water) | |
D-Glutamic acid dimethyl ester hydrochloride |
27025-25-8 | |
MFCD00235872 | |
MFUPLHQOVIUESQ-NUBCRITNSA-N | |
dimethyl (2R)-2-aminopentanedioate;hydrochloride | |
12917567 | |
98% | |
Hygroscopic |
Sicherheit und Handhabung
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only