Learn More
Poulten & Graf GmbH Desalination Unit LAB-IoN L 2
With the desalination unit, Poulten & Graf GmbH Desalination Unit LAB-IoN L 2, demineralised water can be produced in the simplest way.
Brand:  Poulten & Graf GmbH 110 325
 View more versions of this product
Product Code. 10595882
  • 1120.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Used cartridges are simply replaced
  • Connection tube to a 3/4in. tap
  • Includes pH meter
  • Includes mounting
Specifications
Specifications
30 to 40 L/hr.
8 diodes
Desalination Unit