Learn More
Poulten & Graf GmbH Desalination Unit LAB-IoN L 2
With the desalination unit, Poulten & Graf GmbH Desalination Unit LAB-IoN L 2, demineralised water can be produced in the simplest way.
1000.00 € - 1120.00 €
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10478611
|
Poulten & Graf GmbH
110 320 |
4 diodes |
1000.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
10595882
|
Poulten & Graf GmbH
110 325 |
8 diodes |
1120.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
- Used cartridges are simply replaced
- Connection tube to a 3/4in. tap
- Includes pH meter
- Includes mounting
Specifications
30 to 40 L/hr. | |
Desalination Unit |