Learn More
DMP 777, MedChemExpress
MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
Brand: Medchem Express HY-75957
This item is not returnable.
View return policy
Description
DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.Specifications
DMP 777 | |
Off-White | |
C31H40N4O6 | |
L-694458 | |
CCC[C@H](C1=CC2=C(C=C1)OCO2)NC(N3[C@H](C(CC)(C3=O)CC)OC4=CC=C(C=C4)C(N5CCN(CC5)C)=O)=O | |
564.67 | |
Research | |
COVID-19-immunoregulation |
157341-41-8 | |
Research | |
25 mg | |
DMSO : 38.33 mg/mL (67.88 mM; Need ultrasonic) | |
564.67 | |
98.41% | |
Solid |
Research purposes only