missing translation for 'onlineSavingsMsg'
Learn More

DMP 777, MedChemExpress

MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.

265.95 € - 1082.70 €

Specifications

CAS 157341-41-8
Molecular Formula C31H40N4O6
Molecular Weight (g/mol) 564.67
Percent Purity 98.41%
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16490638
Encompass_Preferred
View Documents
Medchem Express
HY-75957
5 mg
265.95 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16484097
Encompass_Preferred
View Documents
Medchem Express
HY-75957
50 mg
1082.70 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16408208
Encompass_Preferred
View Documents
Medchem Express
HY-75957
10 mg
398.25 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16403776
Encompass_Preferred
View Documents
Medchem Express
HY-75957
25 mg
677.70 €
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
Specifications

Specifications

157341-41-8
C31H40N4O6
564.67
98.41%
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)

Recommended Storage : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)

SDS
Documents

Documents

Product Certifications

Research purposes only