Learn More
DMP 777, MedChemExpress
MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
Brand:  Medchem Express HY-75957
Encompass_Preferred
 View more versions of this product
Product Code. 16408208
  • 398.25 € / 10mg
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
This item is not returnable. View return policy
Description
Description
DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
Specifications
Specifications
DMP 777
Off-White
C31H40N4O6
L-694458
CCC[C@H](C1=CC2=C(C=C1)OCO2)NC(N3[C@H](C(CC)(C3=O)CC)OC4=CC=C(C=C4)C(N5CCN(CC5)C)=O)=O
564.67
Research
COVID-19-immunoregulation
157341-41-8
Research
10 mg
DMSO : 38.33 mg/mL (67.88 mM; Need ultrasonic)
564.67
98.41%
Solid